Learn More
Medchemexpress LLC Daclatasvir dihydrochloride | 1009119-65-6 | MFCD25541736 | 99.8% | 811.80 | C40H52Cl2N8O6 | 5 MG
SDP
Catalog No. 5000244185
Encompass_Preferred
Daclatasvir dihydrochloride is the dihydrochloride salt of daclatasvir, a potent inhibitor of hepatitis C virus nonstructural protein 5A (NS5A) supplied for research and analytical use. It is provided as a white to yellow solid and as prepared DMSO stock solutions for in vitro and transporter studies.
  • High purity (99.8%) suitable for research applications.
  • Potent NS5A inhibition with EC50s in the picomolar range across HCV genotypes.
  • Available as dry powder and as 10 mM solutions in DMSO for assays.
  • Recommended storage: sealed at 4°C; in solvent store at -80°C or -20°C per stability guidance.
  • Supplied in small quantities appropriate for assay development and analytical work.
Catalog No. 50-002-44185 Supplier Medchemexpress LLC Supplier No. HY10465R5MG
Daclatasvir dihydrochloride is the dihydrochloride salt of daclatasvir, a potent inhibitor of hepatitis C virus nonstructural protein 5A (NS5A) supplied for research and analytical use. It is provided as a white to yellow solid and as prepared DMSO stock solutions for in vitro and transporter studies.
  • High purity (99.8%) suitable for research applications.
  • Potent NS5A inhibition with EC50s in the picomolar range across HCV genotypes.
  • Available as dry powder and as 10 mM solutions in DMSO for assays.
  • Recommended storage: sealed at 4°C; in solvent store at -80°C or -20°C per stability guidance.
  • Supplied in small quantities appropriate for assay development and analytical work.