Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Daclatasvir dihydrochloride | 1009119-65-6 | MFCD25541736 | 99.8% | 811.80 | C40H52Cl2N8O6 | 50 MG
SDP

Catalog No. 5000241801
Encompass_Preferred

Daclatasvir dihydrochloride is the dihydrochloride salt of daclatasvir, a potent, orally active inhibitor of the hepatitis C virus NS5A protein used for research and analytical applications.

  • Potent HCV NS5A inhibitor with EC50 9-146 pM.
  • High purity (99.82%).
  • Solid form suitable for analytical and research use.
  • Molecular weight 811.80; formula C40H52Cl2N8O6.
  • Inhibits OATP1B and OATP1B3 (IC50 1.5 μM and 3.27 μM).
  • Storage: in solvent -80°C (up to 6 months) or -20°C (up to 1 month) when sealed.

Catalog No. 50-002-41801 Supplier Medchemexpress LLC Supplier No. HY1046550MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Daclatasvir dihydrochloride is the dihydrochloride salt of daclatasvir, a potent, orally active inhibitor of the hepatitis C virus NS5A protein used for research and analytical applications.

  • Potent HCV NS5A inhibitor with EC50 9-146 pM.
  • High purity (99.82%).
  • Solid form suitable for analytical and research use.
  • Molecular weight 811.80; formula C40H52Cl2N8O6.
  • Inhibits OATP1B and OATP1B3 (IC50 1.5 μM and 3.27 μM).
  • Storage: in solvent -80°C (up to 6 months) or -20°C (up to 1 month) when sealed.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.