Learn More
Medchemexpress LLC Daclatasvir dihydrochloride | 1009119-65-6 | MFCD25541736 | 99.8% | 811.80 g/mol | C40H52Cl2N8O6 | 500 MG
SDP
Catalog No. 5000243302
Encompass_Preferred
Daclatasvir dihydrochloride is the dihydrochloride salt of daclatasvir, a potent NS5A inhibitor used in antiviral research against hepatitis C virus (HCV). Supplied as a white-to-yellow solid with high analytical purity, it is intended for laboratory research applications including biochemical and cellular assays.
  • High purity suitable for research-grade applications.
  • Dihydrochloride salt form for improved solubility.
  • White-to-yellow solid powder, easy to weigh and store.
  • Used as an NS5A inhibitor in HCV-related biochemical studies.
  • Characterized with validated identity and molecular weight information.
Catalog No. 50-002-43302 Supplier Medchemexpress LLC Supplier No. HY10465500MG
Daclatasvir dihydrochloride is the dihydrochloride salt of daclatasvir, a potent NS5A inhibitor used in antiviral research against hepatitis C virus (HCV). Supplied as a white-to-yellow solid with high analytical purity, it is intended for laboratory research applications including biochemical and cellular assays.
  • High purity suitable for research-grade applications.
  • Dihydrochloride salt form for improved solubility.
  • White-to-yellow solid powder, easy to weigh and store.
  • Used as an NS5A inhibitor in HCV-related biochemical studies.
  • Characterized with validated identity and molecular weight information.