Learn More
Medchemexpress LLC HY-13272 10mg , Dacomitinib CAS:1110813-31-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1327210MG
Encompass_Preferred
HY-13272 10mg Dacomitinib CAS: 1110813-31-4 Dacomitinib (PF-00299804) is a specific and irreversible inhibitor of the ERBB family of kinases with IC50s of 6 nM, 45.7 nM and 73.7 nM for EGFR, ERBB2, and ERBB4, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9242