Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Dacomitinib | 1110813-31-4 | 469.94 | 200 MG
SDP

Catalog No. 5000366039
Encompass_Preferred

Dacomitinib (PF-00299804) is a specific and irreversible inhibitor of the ERBB family of kinases. It exhibits IC50s of 6 nM, 45.7 nM, and 73.7 nM for EGFR, ERBB2, and ERBB4, respectively.

  • Targets EGFR and apoptosis
  • Inhibits JAK/STAT signaling and protein tyrosine kinase/RTK pathways
  • Available as a solid
  • Molecular formula: C24H25ClFN5O2
  • Recommended storage for powder: -20°C (3 years) or 4°C (2 years)
  • Recommended storage in solvent: -80°C (2 years) or -20°C (1 year)

Catalog No. 50-003-66039 Supplier Medchemexpress LLC Supplier No. 200MGHY13272
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Dacomitinib (PF-00299804) is a specific and irreversible inhibitor of the ERBB family of kinases. It exhibits IC50s of 6 nM, 45.7 nM, and 73.7 nM for EGFR, ERBB2, and ERBB4, respectively.

  • Targets EGFR and apoptosis
  • Inhibits JAK/STAT signaling and protein tyrosine kinase/RTK pathways
  • Available as a solid
  • Molecular formula: C24H25ClFN5O2
  • Recommended storage for powder: -20°C (3 years) or 4°C (2 years)
  • Recommended storage in solvent: -80°C (2 years) or -20°C (1 year)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.