Learn More
Apexbio Technology LLC Dacomitinib (PF299804, PF299) 1110813-31-4 10mM (in 1mL DMSO)
SDP
Catalog No. 5000062514
Encompass_Preferred
Dacomitinib (PF299804 CAS 1110813-31-4) is an irreversible pan-HER small-molecule inhibitor that targets the epidermal growth factor receptor (EGFR/ErbB-1) ERBB2 (HER2) and ERBB4 (HER4) It acts by covalently binding EGFR family tyrosine kinases inhibiting downstream signaling through phospho-HER family proteins AKT and ERK Dacomitinib shows activity in vitro inducing G0/G1 cell cycle arrest and apoptosis and suppresses proliferation in trastuzumab- or lapatinib-resistant HER2-amplified breast cancer cell lines In vivo studies demonstrate antitumor effects in EGFR-mutant (e g T790M) gefitinib-resistant lung cancer xenografts making it relevant for non-small cell lung cancer research
Catalog No. 50-000-62514 Supplier Apexbio Technology LLC Supplier No. A83195.1
Dacomitinib (PF299804 CAS 1110813-31-4) is an irreversible pan-HER small-molecule inhibitor that targets the epidermal growth factor receptor (EGFR/ErbB-1) ERBB2 (HER2) and ERBB4 (HER4) It acts by covalently binding EGFR family tyrosine kinases inhibiting downstream signaling through phospho-HER family proteins AKT and ERK Dacomitinib shows activity in vitro inducing G0/G1 cell cycle arrest and apoptosis and suppresses proliferation in trastuzumab- or lapatinib-resistant HER2-amplified breast cancer cell lines In vivo studies demonstrate antitumor effects in EGFR-mutant (e g T790M) gefitinib-resistant lung cancer xenografts making it relevant for non-small cell lung cancer research