Learn More
Apexbio Technology LLC Dacomitinib (PF299804, PF299), 5mg. Cas: 1110813-31-4 MFCD: MFCD19443734. HER inhibitor
SDP
Catalog No. 501905106
Encompass_Preferred
IC50: 6 nM (EGFR); 45.7 nM (ERBB2); 73.7 nM (ERBB4)The epidermal growth factor receptor (EGFR; ErbB-1; HER1 in humans) is the cell-surface receptor for members of the epidermal growth factor family (EGF-family) of extracellular protein ligands. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5106 Supplier Apexbio Technology LLC Supplier No. A83195
IC50: 6 nM (EGFR); 45.7 nM (ERBB2); 73.7 nM (ERBB4)The epidermal growth factor receptor (EGFR; ErbB-1; HER1 in humans) is the cell-surface receptor for members of the epidermal growth factor family (EGF-family) of extracellular protein ligands. Other sizes are available. Please inqury us for quote.