Learn More
Aviva Systems Biology Corp DACT1 Antibody - N-terminal region (ARP62790_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP62790_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against DACT1. It was validated on Western Blot. . Alternate names: DAPPER, DAPPER1, DPR1, FRODO, HDPR1, THYEX3ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-287-771