Learn More
TARGETMOL CHEMICALS INC Dactolisib 200MG
SDP
Catalog No. 502877562
Encompass_Preferred
Also available in 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Dactolisib (BEZ235) is an orally bioavailable inhibitor of PI3K and mTOR, with IC50 values of 4 nM for p110alpha, 5 nM for p110gamma, 7 nM for p110delta, 75 nM for p110beta, and 20.7 nM for mTOR. Purity 99.71%
Catalog No. 50-287-7562 Supplier TARGETMOL CHEMICALS INC Supplier No. T2235200MG
Also available in 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Dactolisib (BEZ235) is an orally bioavailable inhibitor of PI3K and mTOR, with IC50 values of 4 nM for p110alpha, 5 nM for p110gamma, 7 nM for p110delta, 75 nM for p110beta, and 20.7 nM for mTOR. Purity 99.71%