Learn More
TARGETMOL CHEMICALS INC DACTOLISIB TOSYLATE 25MG
SDP
Catalog No. 502581282
Encompass_Preferred
Also available in 50 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Dactolisib Tosylate (BEZ235 Tosylate) is a dual kinase inhibitor that targets PI3K and mTOR. dactolisib Tosylate exhibits IC50 values of 4 75 7 and 5 nM for PI3K(alpha) (beta) (gamma) and (delta) respectively. dactolisib Tosylate inhibits mTORC1 and mTORC2. purity: 99%
Catalog No. 50-258-1282 Supplier TARGETMOL CHEMICALS INC Supplier No. T1455225MG
Also available in 50 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Dactolisib Tosylate (BEZ235 Tosylate) is a dual kinase inhibitor that targets PI3K and mTOR. dactolisib Tosylate exhibits IC50 values of 4 75 7 and 5 nM for PI3K(alpha) (beta) (gamma) and (delta) respectively. dactolisib Tosylate inhibits mTORC1 and mTORC2. purity: 99%