Learn More
Siemens Healthineers Dade Actin™ Activated Cephaloplastin Reagent
Catalog No. 23044648 Shop All Siemens Healthineers Products
Change view
Click to view available options
Quantity:
10 x 2 mL
10 x 10 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-044-648 10 x 10 mL
23-044-646 10 x 2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 23-044-648 Supplier Siemens Healthineers Supplier No. 10445711
Add to Cart
Add to Cart
This item is not returnable. View return policy
Ideal choice for institutions requiring a moderate screening APTT reagent for routine testing.
Dade Actin™ Activated Cephaloplastin reagent has moderate sensitivity to factor deficiencies (VIII, IX, XI, and XII) in the intrinsic system. It is the ideal choice for institutions requiring a moderate screening APTT reagent for routine testing. Dade Actin™ Activated Cephaloplastin reagent has low heparin sensitivity, allowing the monitoring of heparin therapy even with high heparin dosage. It has moderate sensitivity to lupus anticoagulants.
TRUSTED_SUSTAINABILITY
Specifications
Type Hemostasis Reagent
Quantity 10 x 10 mL