Learn More
Siemens Healthineers Dade Actin™ FSL Activated PTT Reagent
Catalog No. 23044651 Shop All Siemens Healthineers Products
Change view
Click to view available options
Quantity:
10 x 2 mL
10 x 10 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-044-651 10 x 10 mL
23-044-650 10 x 2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 23-044-651 Supplier Siemens Healthineers Supplier No. 10445714
Add to Cart
Add to Cart
This item is not returnable. View return policy
Shows good factor sensitivity to detect clinically significant deficiencies of the intrinsic system.
Dade Actin™ FSL Activated PTT reagent exhibits increased sensitivity to lupus anticoagulants and moderate heparin sensitivity. The reagent shows good factor sensitivity to detect clinically significant deficiencies of the intrinsic system.
TRUSTED_SUSTAINABILITY
Specifications
Type Hemostasis Reagent
Quantity 10 x 10 mL