Learn More
Siemens Healthineers Dade™ Ci-Trol™ Coagulation Controls
Catalog No. 23044662 Shop All Siemens Healthineers Products
Change view
Click to view available options
Level:
1
2
3
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Level
23-044-662 1
23-044-663 2
23-044-664 3
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 23-044-662 Supplier Siemens Healthineers Supplier No. 10445731
Add to Cart
Add to Cart
This item is not returnable. View return policy
For use as control in citrated coagulation systems.
  • Available in Levels 1 to 3.
  • Used for quality control purposes in Prothrombin Time (PT) & Activated Partial Thromboplastin Time (APTT) testing.
TRUSTED_SUSTAINABILITY
Specifications
Type Coagulation Control
For Use With (Application) Prothrombin Time (PT)/Activated Partial Thromboplastin Time (APTT) Testing
Level 1
Quantity 20 x 1 mL