Learn More
Siemens Healthineers Dade™ Data-Fi™ Abnormal Fibrinogen Control
Catalog No. 23044654 Shop All Siemens Healthineers Products
Change view
Click to view available options
Level:
Low
Catalog No. Level
23-044-654 Low
1 options
This item is not returnable. View return policy
Catalog No. 23-044-654 Supplier Siemens Healthineers Supplier No. 10445719
Add to Cart
Add to Cart
This item is not returnable. View return policy
Assessment of accuracy and precision of Dade™ Fibrinogen Determination Reagents in the low range.
Dade™ Data-Fi™ Abnormal Fibrinogen Control Plasma is a control derived from human plasma. It is used to assess accuracy and precision of Dade Fibrinogen Determination Reagents in the low range.
TRUSTED_SUSTAINABILITY
Specifications
Type Hemostasis Reagent
For Use With (Application) Fibrinogen Testing
Level Low