Learn More
Siemens Healthineers Dade Fibrinogen Determination Kit
Catalog No. 23044781 Shop All Siemens Healthineers Products
Change view
Click to view available options
Quantity:
50 Tests/Kit
Catalog No. Quantity
23-044-781 50 Tests/Kit
1 options
This item is not returnable. View return policy
Catalog No. 23-044-781 Supplier Siemens Healthineers Supplier No. 10873571
Add to Cart
Add to Cart
This item is not returnable. View return policy
Quantitative determination of fibrinogen in human plasma.
The Dade Fibrinogen Determination Reagent consists of Dade Thrombin Reagent, Fibrinogen Standard and Dade Owren’s Veronal Buffer for use in the determination of fibrinogen (Clauss method) in the detection of hereditary or acquired hypo- and hyperfibrinogenemia, dysfibrinogenemia, and afibrinogenemia. The reagent offers long stability after reconstitution.
TRUSTED_SUSTAINABILITY
Specifications
Type Hemostasis Kit
For Use With (Application) Fibrinogen Testing
Quantity 50 Tests/Kit
WARNING: Reproductive Harm - www.P65Warnings.ca.gov