Learn More
Siemens Healthineers Dade™ Innovin™ PT Reagent (US)
Catalog No. 23044778 Shop All Siemens Healthineers Products
Change view
Click to view available options
Quantity:
10 x 4 mL
10 x 10 mL
12 x 20 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-044-778 12 x 20 mL
23-044-776 10 x 4 mL
23-044-777 10 x 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 23-044-778 Supplier Siemens Healthineers Supplier No. 10873568
Add to Cart
Add to Cart
This item is not returnable. View return policy
For use in prothrombin time (PT) determinations and prothrombin time-based assays in citrated human plasma.
Dade™ Innovin™ Reagent is prepared from purified recombinant human tissue factor produced in E. coli combined with synthetic phospholipids, calcium, buffers, and stabilizers. The reagent contains minimal residual clotting factors, such as prothrombin and factors VII or X, for clear definition of factor deficiencies and steep factor assay curves. It is highly sensitive to extrinsic factor deficiencies and oral anticoagulant-treated patient plasma samples.
The sensitivity of the Dade™ Innovin™ Reagent is very similar to the WHO human brain reference thromboplastin. It is insensitive to therapeutic levels of heparin, which, in combination with its high sensitivity to coagulation factors, makes Dade™ Innovin™ ideal for monitoring oral anticoagulant therapy and differentiating abnormal plasmas, even in the mildly pathological range.
TRUSTED_SUSTAINABILITY
Specifications
Type Hemostasis Reagent
For Use With (Application) Prothrombin Time (PT) Testing
Quantity 12 x 20 mL