Learn More
Siemens Healthineers Dade Thrombin Reagent
Catalog No. 23044655 Shop All Siemens Healthineers Products
Change view
Click to view available options
Quantity:
10 x 1 mL
10 x 5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-044-655 10 x 1 mL
23-044-656 10 x 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 23-044-655 Supplier Siemens Healthineers Supplier No. 10445720
Add to Cart
Add to Cart
This item is not returnable. View return policy
Determination of fibrinogen in plasma and to accelerate coagulation of anticoagulated samples for immunohematology studies.
Dade Thrombin Reagent is an effective reagent for use in the determination (Clauss method) of fibrinogen in the detection of hereditary or acquired hypo- and hyperfibrinogenemia, dysfibrinogenemia, and afibrinogenemia. The reagent offers long stability after reconstitution.
TRUSTED_SUSTAINABILITY
Specifications
Type Hemostasis Reagent
For Use With (Application) Fibrinogen Testing
Quantity 10 x 1 mL