Learn More
Eurofins Discoverx [D-Ala2, D-Leu5]-Enkephalin (DADLE), 5 mg. (92-1055)
Supplier:  Eurofins Discoverx 921055
Encompass
[D-Ala2, D-Leu5]-Enkephalin (DADLE) is a synthetic peptide that is a selective agonist of the delta opioid receptor, but also binds mu opioid receptors of subtype μ1. It has antinociceptive and analgesic properties via neuron protection, and in-vivo effects include reduction in arterial blood pressure and heart rate. Provided as 5 mg of dry powder alongside reconstitution buffer. These highly purified, pre-validated control ligands have been used to qualify our InCELL Hunter,  PathHunter and cAMP Hunter cell lines as well as our Assay Ready Kits. Ligands are conveniently supplied with a recommended reconstitution buffer. For detailed information and product-specific datasheet, visit www.discoverx.com/ligands.
Catalog No. 50-199-9596