Learn More
Abcam DADLE, delta-Opioid peptide agonist, 1MG
Catalog No. 502707364
Encompass_Preferred
MW 569.66 g/mol, Purity >95%. Synthetic δ-opioid peptide agonist, with some activity at μ-opioid receptors. Neuro- and tissue-protective properties. Attenuates mRNA expression of p53 and c-fos.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7364 Supplier Abcam Supplier No. AB1206731MG
MW 569.66 g/mol, Purity >95%. Synthetic δ-opioid peptide agonist, with some activity at μ-opioid receptors. Neuro- and tissue-protective properties. Attenuates mRNA expression of p53 and c-fos.
The product is subject to the following: Abcam Restricted Use Statement