Learn More
AdipoGen DAF-2
SDP
Supplier:  AdipoGen CDXD0084M005
Encompass_Preferred
Chemical. CAS 205391-01-1. Formula C20H14N2O5. MW 362.34. Synthetic. DAF-2 is highly sensitive reagent for NO detection and determination of nitric oxide synthase activity. DAF-2, however, remains essentially nonfluorescent until it reacts with the nitrosonium cation produced by spontaneous oxidation of nitric oxide to form a fluorescent heterocycle, which becomes trapped in the cells cytoplasm. This sensitive fluorescent probe has been used to identify individual nitric oxide-producing neurons in brain slices, in mitochondria and in living plant cells. Spectral data lambdaex 505nm lambdaem 518nm in 0.1M Tris pH8.0.
Catalog No. 50-256-1254