Learn More
AdipoGen DAF-2 DA
SDP
Supplier:  AdipoGen CDXD0085M010
Encompass_Preferred
Chemical. CAS 205391-02-2. Formula C24H18N2O7. MW 446.41. Synthetic. DAF-2 DA is probably the most successful indicator for nitric oxide. As with other fluorescein diacetates, DAF-2 diacetate is membrane permeant and is deacetylated by intracellular esterases to 4,5-diaminofluorescein DAF-2. Can be used for the fluorimetric detection of nitric oxide and in fluorescence microscopy to measure real-time changes in nitric oxide levels in vivo. Spectral Data lambdaex 491nm lambdaem 513nm.
Catalog No. 50-256-1251