Learn More
Enzo Life Sciences DAF-2 DA (cell permeable) (1mg). CAS: 205391-02-2
SDP
Catalog No. 502009600
Encompass_Preferred
Cell permeable analog of DAF-2 (ALX-620-052) that is hydrolyzed to DAF-2 by intracellular esterases. Can be used in fluorescence microscopy to measure real-time changes in nitric oxide (NO) levels in vivo (Ex(max): 495nm; Em(max): 515nm). Alternative name: 4,5-Diaminofluorescein . diacetate. Formulation: Dissolved in 0.45ml DMSO. Purity: ≥98% (HPLC). Long Term Storage: -20°C.
Catalog No. 50-200-9600 Supplier Enzo Life Sciences Supplier No. ALX620056M001
Cell permeable analog of DAF-2 (ALX-620-052) that is hydrolyzed to DAF-2 by intracellular esterases. Can be used in fluorescence microscopy to measure real-time changes in nitric oxide (NO) levels in vivo (Ex(max): 495nm; Em(max): 515nm). Alternative name: 4,5-Diaminofluorescein . diacetate. Formulation: Dissolved in 0.45ml DMSO. Purity: ≥98% (HPLC). Long Term Storage: -20°C.