Learn More
Abcam DAF-2 DA (DMSO solution), fluorescent nitric o x ide probe, 100UG
Catalog No. 502704543
Encompass_Preferred
MW 446.4 Da, Purity >98%. Fluorescent nitric oxide probe; membrane-permeable. Once inside cell it reacts with NO in the presence of oxygen to yield the highly fluorescent triazolofluorescein DAF-2T (ab145473). Determines nitric oxide synthase activity. Has a 2-5 nM detection limit at neutral pH. Also available as a solid (ab145283).
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-4543 Supplier Abcam Supplier No. AB146631100UG
MW 446.4 Da, Purity >98%. Fluorescent nitric oxide probe; membrane-permeable. Once inside cell it reacts with NO in the presence of oxygen to yield the highly fluorescent triazolofluorescein DAF-2T (ab145473). Determines nitric oxide synthase activity. Has a 2-5 nM detection limit at neutral pH. Also available as a solid (ab145283).
The product is subject to the following: Abcam Restricted Use Statement