Learn More
Cayman Chemical DAF-2 dIacetate 100UG
SDP
Catalog No. 502896681
Encompass_Preferred
DAF-2 diacetate is a sensitive fluorescent indicator for the detection and bioimaging of NO.{6360,6632} It is a cell-permeable derivative of DAF-2. Upon entry into the cell, DAF-2 diacetate is transformed into the less cell-permeable DAF-2 by cellular esterases thus preventing loss of signal due to diffusion of the molecule from the cell. In the presence of oxygen, DAF-2 reacts with NO to yield the highly fluorescent triazolofluorescein (DAF-2T). Fluorescence is monitored using excitation and emission wavelengths of 485 and 538 nm, respectively.{6632} At neutral pH the detection limit for NO is 2-5 nM. DAF-2 diacetate can be utilized in cells which produce small amounts of NO, such as endothelial cells, as well as in cells which generate large amount of NO, such as macrophages.{6360,6632}
Catalog No. 50-289-6681 Supplier Cayman Chemical Supplier No. 85165100 UG
DAF-2 diacetate is a sensitive fluorescent indicator for the detection and bioimaging of NO.{6360,6632} It is a cell-permeable derivative of DAF-2. Upon entry into the cell, DAF-2 diacetate is transformed into the less cell-permeable DAF-2 by cellular esterases thus preventing loss of signal due to diffusion of the molecule from the cell. In the presence of oxygen, DAF-2 reacts with NO to yield the highly fluorescent triazolofluorescein (DAF-2T). Fluorescence is monitored using excitation and emission wavelengths of 485 and 538 nm, respectively.{6632} At neutral pH the detection limit for NO is 2-5 nM. DAF-2 diacetate can be utilized in cells which produce small amounts of NO, such as endothelial cells, as well as in cells which generate large amount of NO, such as macrophages.{6360,6632}