Learn More
Abcam DAF-2, Fluorescent nitric o x ide probe, 1MG
Catalog No. 502705349
Encompass_Preferred
MW 362.34 g/mol, Purity >95%. Highly sensitive, specific fluorescent detector for nitric oxide (NO). Essentially nonfluorescent until it reacts with NO to yield the highly fluorescent triazolofluorescein (DAF-2T) (ab145473).
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5349 Supplier Abcam Supplier No. AB1452691MG
MW 362.34 g/mol, Purity >95%. Highly sensitive, specific fluorescent detector for nitric oxide (NO). Essentially nonfluorescent until it reacts with NO to yield the highly fluorescent triazolofluorescein (DAF-2T) (ab145473).
The product is subject to the following: Abcam Restricted Use Statement