Learn More
AdipoGen DAF-2T
SDP
Supplier:  AdipoGen CDXD0044M001
Encompass_Preferred
Chemical. CAS 208850-35-5. Formula C20H11N3O5. MW 373.32. Synthetic. Reference material for DAF-2 CDX-D0084. Spectral data lambdaex 495nm lambdaem 515nm.
Catalog No. 50-256-1287