Learn More
AdipoGen DAF-4 DA
SDP
Supplier:  AdipoGen CDXD0216M010
Encompass_Preferred
Chemical. CAS 208850-34-4 parent compound DAF-4. Formula C24H16Cl2N2O7. MW 515.3. Synthetic. DAF-4 is a indicator for nitric oxide. As with other fluorescein diacetates DAFs, the non-fluorescent DAF-4 diacetate DAF-4 DA is membrane permeant and is deacetylated by intracellular esterases to form DAF-4, which than can ract with NO to form the fluorescent DAF-4T. Can be used for the fluorimetric detection of nitric oxide and in fluorescence microscopy to measure real-time changes in nitric oxide levels in vivo. Spectral Data lambdaex 505nm lambdaem 530nm.
Catalog No. 50-256-1176