Learn More
AdipoGen DAF-4T (5 mg)
SDP
Catalog No. 502054373
Encompass_Preferred
Chemical. CAS: 212558-80-0. Formula: C20H7O5N3Cl2. MW: 440.19. Fluorescent reference compound for DAF-4 DA (CDX-D0216). Spectral data: lambdaEx=505nm; lambdaEm=530nm.
Catalog No. 50-205-4373 Supplier AdipoGen Supplier No. CDXD0211M005
Chemical. CAS: 212558-80-0. Formula: C20H7O5N3Cl2. MW: 440.19. Fluorescent reference compound for DAF-4 DA (CDX-D0216). Spectral data: lambdaEx=505nm; lambdaEm=530nm.