Learn More
AdipoGen DAF-4T Solution 5 mM in DMSO
SDP
Catalog No. 502561007
Encompass_Preferred
Chemical. CAS 212558-80-0. Formula C20H7Cl2N3O5. MW 440.19. Fluorescent reference compound for DAF-4 DA CDX-D0216. Spectral data lambdaEx=505nm lambdaEm=530nm.
Catalog No. 50-256-1007 Supplier AdipoGen Supplier No. CDXD1233M001
Chemical. CAS 212558-80-0. Formula C20H7Cl2N3O5. MW 440.19. Fluorescent reference compound for DAF-4 DA CDX-D0216. Spectral data lambdaEx=505nm lambdaEm=530nm.