Learn More
AdipoGen DAF-FM
SDP
Supplier:  AdipoGen CDXA0023M001
Encompass_Preferred
Chemical. CAS 254109-20-1. Formula C21H14F2N2O5. MW 412.34. Synthetic. DAF-FM is an important reagent for quantitating low concentrations of nitric oxide in solution. This compound is essentially nonfluorescent until it reacts with NO to form a fluorescent benzotrizole. With excitation/emission maxima of 495/515 nm, DAF-FM can be detected using any instrument that can be used to detect fluorescein.
Catalog No. 50-256-1861