Learn More
AdipoGen DAF-FM DA
SDP
Supplier:  AdipoGen CDXA0024M005
Encompass_Preferred
Chemical. CAS 254109-22-3. Formula C25H18F2N2O7. MW 496.42. Synthetic. DAF-FM diacetate is an important reagent for quantification of low concentrations of nitric oxide in solution. This compound is essentially nonfluorescent until it reacts with NO to form a fluorescent benzotriazole. With excitation/emission maxima of 495/515 nm, DAF-FM can be detected using any instrument suitable for detecting fluorescein.
Catalog No. 50-256-1858