Learn More
Enzo Life Sciences DAF-FM DA (cell permeable) (1mg). CAS: 254109-22-3
SDP
Supplier:  Enzo Life Sciences ALX620071M001
Encompass_Preferred
Cell permeable photo-stable nitric oxide (NO) fluorescent indicator, that is hydrolyzed to DAF-FM by intracellular esterases (Ex: 500nm; Em: 515nm). Stable in a pH range of 5.5-12. High yield (quantum efficiency). Detection limit 3nM. Not for sale in Japan. Alternative name: Diaminofluorescein-FM . diacetate. Formulation: Dissolved in 0.4ml DMSO. Purity: ≥98% (HPLC). Long Term Storage: -20°C.
Catalog No. 50-200-9603