Learn More
Cayman Chemical DAF-FM diacetate, Purity: ≥98%, CAS: 254109-22-3, 1 mg
SDP
Catalog No. 502979939
Encompass_Preferred
A cell-permeable, fluorescent probe for the detection and bioimaging of NO (ex. max 495 nm/em. max 515 nm)
Catalog No. 50-297-9939 Supplier Cayman Chemical Supplier No. 187671 MG
A cell-permeable, fluorescent probe for the detection and bioimaging of NO (ex. max 495 nm/em. max 515 nm)