Learn More
Aviva Systems Biology Corp Dag1 Antibody - C-terminal region (ARP46446_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP46446_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Dag1. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: D9Wsu13e, DG, Dp427, Dp71ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-285-259