Learn More
Sino Biological DAG1/Dystroglycan Antibody, Rabbit PAb, Antigen Affinity Purified 100uL
Supplier:  Sino Biological 14421T24100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human DAG1/Dystroglycan (rh DAG1/Dystroglycan; Catalog#14421-H08H; NP_004384.4; Met1-Val749). DAG1/Dystroglycan specific IgG was purified by Human DAG1/Dystroglycan affinity chromatography.
Catalog No. 50-224-8372