Learn More
Aviva Systems Biology Corp DAGLB Antibody - middle region (ARP55430_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP55430_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against DAGLB. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human DAGLB. Alternate names: DAGLBETA, FLJ33624, FLJ33909, KCCR13LApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-53754