Learn More
Medchemexpress LLC Dagrocorat | 1044535-52-5 | 99.9% | 494.55 | 50 MG
SDP
Catalog No. 5000391171
Encompass_Preferred
Dagrocorat (PF-00251802) is an orally active and selective high-affinity partial agonist of the glucocorticoid receptor. It is also a time-dependent reversible inhibitor of CYP3A (IC50=1.3 μM in human liver microsomes) and CYP2D6 (Ki=0.57 μM in human liver microsomes). It can be used for the research of rheumatoid arthritis.
  • Orally active and selective high-affinity partial agonist of the glucocorticoid receptor.
  • Time-dependent reversible inhibitor of CYP3A (IC50=1.3 μM in human liver microsomes) and CYP2D6 (Ki=0.57 μM in human liver microsomes).
  • Can be used for the research of rheumatoid arthritis.
Catalog No. 50-003-91171 Supplier Medchemexpress LLC Supplier No. HY1671850MG
Dagrocorat (PF-00251802) is an orally active and selective high-affinity partial agonist of the glucocorticoid receptor. It is also a time-dependent reversible inhibitor of CYP3A (IC50=1.3 μM in human liver microsomes) and CYP2D6 (Ki=0.57 μM in human liver microsomes). It can be used for the research of rheumatoid arthritis.
  • Orally active and selective high-affinity partial agonist of the glucocorticoid receptor.
  • Time-dependent reversible inhibitor of CYP3A (IC50=1.3 μM in human liver microsomes) and CYP2D6 (Ki=0.57 μM in human liver microsomes).
  • Can be used for the research of rheumatoid arthritis.