Learn More
Medchemexpress LLC Dagrocorat hydrochloride | 1044535-61-6 | 99.9% | 531.01 | 25 MG
SDP
Catalog No. 5000392028
Encompass_Preferred
Dagrocorat hydrochloride is an orally active and selective high-affinity partial agonist of the glucocorticoid receptor. It is also a time-dependent reversible inhibitor of CYP3A and CYP2D6. This compound can be used for the research of rheumatoid arthritis, with studies indicating its metabolism by cytochrome P450 (CYP)3A to an N-oxide metabolite, and its role as a reversible inhibitor of several CYPs.
  • Orally active and selective high-affinity partial agonist of the glucocorticoid receptor.
  • Time-dependent reversible inhibitor of CYP3A and CYP2D6.
  • Suitable for research on rheumatoid arthritis.
  • Metabolized by cytochrome P450 (CYP)3A.
  • Reversible inhibitor of several CYPs.
Catalog No. 50-003-92028 Supplier Medchemexpress LLC Supplier No. 25MGHY16718A
Dagrocorat hydrochloride is an orally active and selective high-affinity partial agonist of the glucocorticoid receptor. It is also a time-dependent reversible inhibitor of CYP3A and CYP2D6. This compound can be used for the research of rheumatoid arthritis, with studies indicating its metabolism by cytochrome P450 (CYP)3A to an N-oxide metabolite, and its role as a reversible inhibitor of several CYPs.
  • Orally active and selective high-affinity partial agonist of the glucocorticoid receptor.
  • Time-dependent reversible inhibitor of CYP3A and CYP2D6.
  • Suitable for research on rheumatoid arthritis.
  • Metabolized by cytochrome P450 (CYP)3A.
  • Reversible inhibitor of several CYPs.