Learn More
Research Products International Corp Daidzein [4',7-Dihydroxyisoflavone], 25 Milligrams
SDP
Catalog No. 50213276
Encompass_Preferred
Inactive analog of Genistein. Inhibits casein kinase II activity.
Catalog No. 50-213-276 Supplier Research Products International Corp Supplier No. D200100.025
Inactive analog of Genistein. Inhibits casein kinase II activity.