Learn More
Selleck Chemical LLC Daidzein S1849-1g
SDP
Catalog No. 503108580
Encompass_Preferred
Daidzein is a natural isoflavone phytoestrogen found in Leguminosae used as a component of foods and dietary supplements Daidzein is a soy isoflavone which acts as a PPAR activator
Catalog No. 50-310-8580 Supplier Selleck Chemical LLC Supplier No. S18491G
Daidzein is a natural isoflavone phytoestrogen found in Leguminosae used as a component of foods and dietary supplements Daidzein is a soy isoflavone which acts as a PPAR activator