Learn More
Selleck Chemical LLC Daidzein S1849-50mg
SDP
Catalog No. 503110593
Encompass_Preferred
Daidzein is a natural isoflavone phytoestrogen found in Leguminosae used as a component of foods and dietary supplements Daidzein is a soy isoflavone which acts as a PPAR activator
Catalog No. 50-311-0593 Supplier Selleck Chemical LLC Supplier No. S184950MG
Daidzein is a natural isoflavone phytoestrogen found in Leguminosae used as a component of foods and dietary supplements Daidzein is a soy isoflavone which acts as a PPAR activator