Learn More
Medchemexpress LLC HY-N0019 500mg Medchemexpress, Daidzein CAS:486-66-8 Purity:>98%
SDP
Catalog No. 502082770
Encompass_Preferred
Medchemexpress, HY-N0019 500mg Daidzein CAS:486-66-8 Daidzein is a soy isoflavone, which acts as a PPAR activator. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2770 Supplier Medchemexpress LLC Supplier No. HYN0019500MG
Medchemexpress, HY-N0019 500mg Daidzein CAS:486-66-8 Daidzein is a soy isoflavone, which acts as a PPAR activator. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.