Learn More
Sigma Aldrich Fine Chemicals Biosciences Daidzein 98 synthetic25MG
Catalog No. 502899520
Encompass
Daidzeinnbspis an isoflavone present in soy. Itnbspregulates oxidative stress angiogenesis cell growth and apoptosis. Daidzein has antioxidant antiestrogenic and antiatherogenic properties. It provides protection against osteoporosis.
Catalog No. 50-289-9520 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. D780225MG
Daidzeinnbspis an isoflavone present in soy. Itnbspregulates oxidative stress angiogenesis cell growth and apoptosis. Daidzein has antioxidant antiestrogenic and antiatherogenic properties. It provides protection against osteoporosis.