Learn More
Medchemexpress LLC Daidzein-d6 | 291759-05-2 | 96.1% | 260.27 | 1 MG
SDP
Catalog No. 5000431633
Encompass_Preferred
Daidzein-d6 is a deuterium labeled Daidzein, a soy isoflavone that acts as a PPAR activator. It can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Stable heavy isotopes have been incorporated into drug molecules largely as tracers for quantitation during drug development. Deuteration has potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Acts as a PPAR activator
  • Suitable for use as a tracer
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Features stable heavy isotopes incorporated into the molecule
  • Potential to affect pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-004-31633 Supplier Medchemexpress LLC Supplier No. HYN0019S11MG
Daidzein-d6 is a deuterium labeled Daidzein, a soy isoflavone that acts as a PPAR activator. It can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Stable heavy isotopes have been incorporated into drug molecules largely as tracers for quantitation during drug development. Deuteration has potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Acts as a PPAR activator
  • Suitable for use as a tracer
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Features stable heavy isotopes incorporated into the molecule
  • Potential to affect pharmacokinetic and metabolic profiles of drugs