Learn More
Cayman Chemical Daidzein, Purity: ≥95%, CAS: 486-66-8, 500 mg
SDP
Catalog No. 502887954
Encompass_Preferred
An isoflavone; binds to ERβ (Ki = 2.8 µM) but not ERα up to 1 mM; estrogenic in vitro, increasing ERE-mediated gene transcription via ERβ (EC50 = 2.8 µM); an inhibitor of CAVII and CAXII (Kis = 4.2 and 56 nM, respectively) and selective for them over CAI, II, and IV (Kis = >10,000, >10,000, and 718.7 nM, respectively); reduces tumor growth in a PC3 prostate cancer mouse orthtopic model at 50 mg/kg per day
Catalog No. 50-288-7954 Supplier Cayman Chemical Supplier No. 10005166500 MG
An isoflavone; binds to ERβ (Ki = 2.8 µM) but not ERα up to 1 mM; estrogenic in vitro, increasing ERE-mediated gene transcription via ERβ (EC50 = 2.8 µM); an inhibitor of CAVII and CAXII (Kis = 4.2 and 56 nM, respectively) and selective for them over CAI, II, and IV (Kis = >10,000, >10,000, and 718.7 nM, respectively); reduces tumor growth in a PC3 prostate cancer mouse orthtopic model at 50 mg/kg per day