Learn More
Abcam Daidzein, Phytoestrogen, 50MG
Catalog No. 502707619
Encompass_Preferred
MW 254.24 Da, Purity >99%. Phytoestrogen, and soy isoflavone. Analog of genistein (ab120112). Various biological actions, including a weak estrogenic / antiestrogenic effect by binding to the nuclear estrogen receptor.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7619 Supplier Abcam Supplier No. AB12039150MG
MW 254.24 Da, Purity >99%. Phytoestrogen, and soy isoflavone. Analog of genistein (ab120112). Various biological actions, including a weak estrogenic / antiestrogenic effect by binding to the nuclear estrogen receptor.
The product is subject to the following: Abcam Restricted Use Statement