Learn More
Selleck Chemical LLC Daidzin S2289-200mg
SDP
Catalog No. 503109555
Encompass_Preferred
Daidzin (Daidzoside Daidzein 7-O-glucoside Daidzein 7-glucoside) a natural organic compound in the class of phytochemicals known as isoflavones is a potent and selective inhibitor of human mitochondrial aldehyde dehydrogenase and inhibits ALDH-I selectively (Ki 20 nM) at least 500 times less effective against ALDH- the cytosolic isozyme (Ki 10 M)
Catalog No. 50-310-9555 Supplier Selleck Chemical LLC Supplier No. S2289200MG
Daidzin (Daidzoside Daidzein 7-O-glucoside Daidzein 7-glucoside) a natural organic compound in the class of phytochemicals known as isoflavones is a potent and selective inhibitor of human mitochondrial aldehyde dehydrogenase and inhibits ALDH-I selectively (Ki 20 nM) at least 500 times less effective against ALDH- the cytosolic isozyme (Ki 10 M)