Learn More
Apexbio Technology LLC DAIDZIN-20MG
SDP
Catalog No. 5000567634
Encompass_Preferred
Daidzin (CAS No 552-66-9) is a naturally occurring isoflavone glycoside found predominantly in soybean and pueraria plants It is extensively studied in the field of biomedical research for its potential therapeutic effects The compound primarily acts by inhibiting aldehyde dehydrogenase a key enzyme in alcohol metabolism which may contribute to its potential in treating alcohol dependency In vitro studies have shown that Daidzin exhibits activity in the nanomolar to low micromolar range indicating strong efficacy at low concentrations Experimental applications of Daidzin include use in various cell models to study its biochemical pathways and effects as well as in animal studies to evaluate its pharmacokinetics and therapeutic potential Additionally Daidzin is utilized in drug screening processes to explore its efficacy and safety as a potential pharmaceutical agent Experimental concentrations typically depend on the specific research design
Catalog No. 50-005-67634 Supplier Apexbio Technology LLC Supplier No. N180720
Daidzin (CAS No 552-66-9) is a naturally occurring isoflavone glycoside found predominantly in soybean and pueraria plants It is extensively studied in the field of biomedical research for its potential therapeutic effects The compound primarily acts by inhibiting aldehyde dehydrogenase a key enzyme in alcohol metabolism which may contribute to its potential in treating alcohol dependency In vitro studies have shown that Daidzin exhibits activity in the nanomolar to low micromolar range indicating strong efficacy at low concentrations Experimental applications of Daidzin include use in various cell models to study its biochemical pathways and effects as well as in animal studies to evaluate its pharmacokinetics and therapeutic potential Additionally Daidzin is utilized in drug screening processes to explore its efficacy and safety as a potential pharmaceutical agent Experimental concentrations typically depend on the specific research design