Learn More
TARGETMOL CHEMICALS INC Daidzin 500MG
SDP
Catalog No. 502879918
Encompass_Preferred
Also available in 1 mL, 10 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Daidzin (Daidzoside) is an isoflavone isolated from soya bean with anti-oxidant, anti-carcinogenic, and anti-atherosclerotic activities. Purity 99.43%
Catalog No. 50-287-9918 Supplier TARGETMOL CHEMICALS INC Supplier No. T2901500MG
Also available in 1 mL, 10 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Daidzin (Daidzoside) is an isoflavone isolated from soya bean with anti-oxidant, anti-carcinogenic, and anti-atherosclerotic activities. Purity 99.43%