Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Daidzin (daidzein 7-O-glucoside) | 552-66-9 | MFCD00017466 | 99.8% | 416.38 | C21H20O9 | 10 MG
SDP

Catalog No. 5000297712
Encompass_Preferred

Daidzin is an isoflavone research standard and a potent, selective inhibitor of mitochondrial aldehyde dehydrogenase-2 (ALDH-2). It displays antioxidant, anticancer, and antiatherosclerotic activities and is used in biochemical and pharmacological research related to ALDH-2 pathways and ethanol metabolism.

  • High purity (99.77% LCMS).
  • White to off-white solid.
  • Molecular weight 416.38 g/mol.
  • CAS number 552-66-9.
  • Recommended storage: powder -20°C (3 years) or 4°C (2 years).

Catalog No. 50-002-97712 Supplier Medchemexpress LLC Supplier No. HYN001810MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Daidzin is an isoflavone research standard and a potent, selective inhibitor of mitochondrial aldehyde dehydrogenase-2 (ALDH-2). It displays antioxidant, anticancer, and antiatherosclerotic activities and is used in biochemical and pharmacological research related to ALDH-2 pathways and ethanol metabolism.

  • High purity (99.77% LCMS).
  • White to off-white solid.
  • Molecular weight 416.38 g/mol.
  • CAS number 552-66-9.
  • Recommended storage: powder -20°C (3 years) or 4°C (2 years).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.